Anti-PRM1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAH308HU01
MSPP-PAH308HU1EA 350 USD
MSPP-PAH308HU1
Anti-PRM1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Protamine 1 (PRM1), derived from recombinant PRM1 (Ala2~His51), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Protamines are small, arginine-rich, nuclear proteins that replace histones late in the haploid phase of spermatogenesis and are believed essential for sperm head condensation and DNA stabilization. Mice, humans, and certain fish have 2 or more different protamines, whereas the sperm of bull, boar, rat, rabbit, guinea pig, and ram have one form of protamine. The 2 human protamines are denoted P1 and P2 (PRM2)., PRM1 and PRM2 have a single intron, consisting of 91 and 163 bp, respectively. Both genes were found to contain typical TATAA and CAAT boxes at conventional distances from the transcription start points which, by use of primary extension experiments, were assigned to nucleotides -91 and -110 for PRM1 and PRM2, respectively.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR