Anti-SULF2 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAH107HU01
MSPP-PAH107HU1EA 350 USD
MSPP-PAH107HU1
Anti-SULF2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Sulfatase 2 (SULF2), derived from recombinant SULF2 (Phe337~Arg649), is reactive with Human/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Sulfatase 2contains an N-terminal signal sequence, followed by a sulfatase domain, a hydrophilic region, and a C-terminal substrate recognition domain.SULF2 shares about 64% identity with SULF1 and 94% identity with mouse Sulf2. PCR detected SULF2 expression in most human tissues examined, with highest levels in ovary, skeletal muscle, stomach, brain, uterus, heart, kidney, and placenta. Western blot analysis of transfected Chinese hamster ovary (CHO) cells and culture medium detected SULF2 proteins with apparent molecular masses of 132, 64, and 60 kDa in the culture medium. Treatment with furin inhibitors resulted in a single 132-kDa species, and treatment with N-glycanase reduced the molecular mass from 132 to 100 kDa.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: SULF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR