Anti-STS Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAG609HU01
MSPP-PAG609HU1EA 350 USD
MSPP-PAG609HU1
Anti-STS Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Steroid Sulfatase Isozyme S (STS), derived from recombinant STS(His235~Arg583), is reactive with Human/Mouse/Horse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

STS catalyzes the conversion of sulfated steroid precursors to estrogens during pregnancy. The encoded protein is found in the endoplasmic reticulum, where it acts as a homodimer.,The deduced 583-residue protein has a molecular mass of 63 kD and contains a 21- or 23-residue signal peptide, 4 possible N-linked glycosylation sites, and 2 potential membrane-spanning domains. Expression in hamster kidney cells (BHK-21) showed localization predominantly in the endoplasmic reticulum, with smaller fractions found in the Golgi, at the cell surface, and in endosomes and lysosomes. The major transcription start site was at position -221 with respect to the AUG translation initiation codon, and the amino acids were renumbered to make the initiating methionine +1.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: STS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR