Anti-LPCAT3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAG531HU01
MSPP-PAG531HU1EA 350 USD
MSPP-PAG531HU1
Anti-LPCAT3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Lysophosphatidylcholine Acyltransferase 3 (LPCAT3), derived from recombinant LPCAT3 (Met122~Arg233), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The deduced 487-amino acid human and mouse MBOAT5 proteins contain 11 predicted transmembrane domains and share 88% amino acid identity. Human MBOAT5 shares 35% amino acid identity with Drosophila nessy.The deduced protein contains 10 putative transmembrane domains and a C-terminal endoplasmic reticulum (ER) retention motif (KKxx). RT-PCR detected ubiquitous Lpcat3 expression, with highest level in testis. Lpcat3 colocalized with an ER marker protein in transfected Chinese hamster ovary cells., Knockdown of Lpcat3 in mouse melanoma cells with small interfering RNA significantly reduced cellular acyltransferase activity toward endogenous lysophosphatidylcholine, lysophosphatidylethanolamine, and lysophosphatidylserine.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: LPCAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR