Anti-SCGB3A1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAG495HU01
MSPP-PAG495HU1EA 350 USD
MSPP-PAG495HU1
Anti-SCGB3A1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Secretoglobin Family 3A, Member 1(SCGB3A1), derived from recombinant SCGB3A1(Phe21~Gly104), is reactive with Human/Pig.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Breast cancer progresses from atypical hyperproliferation to in situ and then invasive carcinomas before culminating in metastatic disease. Ductal carcinoma in situ (DCIS) is thought to be the precursor of invasive ductal carcinoma. The deduced 104-amino acid HIN1 protein contains a signal peptide and is conserved in several species. HIN1 was highly expressed in breast tissue from a woman in the twenty-fifth week of pregnancy. In situ hybridization analysis confirmed the expression of HIN1 in small ducts and lobules, irrespective of their proliferation and hormone receptor status. HIN1 was also highly expressed in other tissues with branching ductal epithelia. No expression was detected in large or DCIS ducts. Western blot analysis showed expression of an 8-kDa protein.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: SCGB3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR