Anti-MAPRE1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAG491HU01
MSPP-PAG491HU1EA 350 USD
MSPP-PAG491HU1
Anti-MAPRE1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Microtubule Associated Protein RP/EB Family, Member 1 (MAPRE1), derived from recombinant MAPRE1 (Ala2~Tyr268), is reactive with Human/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

MAPRE1 is a regulator of microtubule dynamics that localizes at both the growing plus ends of microtubules and the centrosome. It is involved in a variety of cellular processes, including establishment and maintenance of cell polarity, search and capture of chromosomes during mitosis, and positioning of the mitotic spindle during asymmetric cell division., This microtubule localization was abolished in the presence of the microtubule-destabilizing drug nocodazole; upon drug removal, the microtubule distribution of EB1 was recovered and EB1 fluorescence concentrated at the microtubule-organizing center. These results suggested that EB1 is associated with the microtubule network and may be involved in microtubule polymerization and spindle function.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MAPRE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR