Anti-DSC2 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAG436HU01
MSPP-PAG436HU1EA 350 USD
MSPP-PAG436HU1
Anti-DSC2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Desmocollin 2 (DSC2), derived from recombinant DSC2(Arg136~Cys471), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Desmocollin-2 is a calcium-dependent glycoprotein that is a member of the desmocollin subfamily of the cadherin superfamily. These desmosomal family members, along with the desmogleins, are found primarily in epithelial cells where they constitute the adhesive proteins of the desmosome cell-cell junction and are required for cell adhesion and desmosome formation. The desmosomal family members are arranged in two clusters on chromosome 18, occupying less than 650 kb combined. Alternative splicing results in two transcript variants encoding distinct isoforms.Exon 16 is alternatively spliced, giving rise to the a and b forms of the protein. A remarkable degree of conservation of intron position with other cadherins was observed.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: DSC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR