Anti-DSC1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAG435HU02
MSPP-PAG435HU2EA 360 USD
MSPP-PAG435HU2
Anti-DSC1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Desmocollin 1 (DSC1), derived from recombinant DSC1 (Cys30~Ser353), is reactive with Human/Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Desmocollin-1 is a calcium-dependent glycoprotein that is a member of the desmocollin subfamily of the cadherin superfamily. These desmosomal family members, along with the desmogleins, are found primarily in epithelial cells where they constitute the adhesive proteins of the desmosome cell-cell junction and are required for cell adhesion and desmosome formation. The desmosomal family members are arranged in two clusters on chromosome 18, occupying less than 650 kb combined. Alternative splicing results in two transcript variants encoding distinct isoforms. Strongly expressed in epidermis, less in lymph node and tongue. The glycoproteins of 115 kD and 100 kD were named desmocollins to denote that they are involved in the adhesive function of the desmosomes.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: DSC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR