Anti-DLG4 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAG168MU01
MSPP-PAG168MU1EA 360 USD
MSPP-PAG168MU1
Anti-DLG4 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Discs, Large Homolog 4 (DLG4), derived from recombinant DLG4 (Met1~Ser393), is reactive with Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Disks large homolog 4 is a member of the membrane-associated guanylate kinase (MAGUK) family. It heteromultimerizes with DLG2. With DLG2 it is recruited into the same NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins. DLG4 is the best studied member of the MAGUK-family of PDZ domain-containing proteins. Like all MAGUK-family proteins, its basic structure includes three PDZ domains, an SH3 domain, and a guanylate kinase-like domain (GK) connected by disordered linker regions. It is almost exclusively located in the post synaptic density of neurons, and is involved in anchoring synaptic proteins.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: DLG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR