Anti-TRPV1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAF839HU01
MSPP-PAF839HU1EA 350 USD
MSPP-PAF839HU1
Anti-TRPV1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Transient Receptor Potential Cation Channel Subfamily V, Member 1 (TRPV1), derived from recombinant TRPV1 (Met1~Glu153), is reactive with Human/Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

TRPV1 is a nonselective cation channel that may be activated by a wide variety of exogenous and endogenous physical and chemical stimuli. The best-known activators of TRPV1 are heat greater than 43 °C and capsaicin, the pungent compound in hot chili peppers. The activation of TRPV1 leads to painful, burning sensation. Its endogenous activators include: low pH, the endocannabinoid anandamide, N-arachidonoyl-dopamine. TRPV1 receptors are found mainly in the nociceptive neurons of the peripheral nervous system, but they have also been described in many other tissues, including the central nervous system. TRPV1 is involved in the transmission and modulation of pain, as well as the integration of diverse painful stimuli.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: TRPV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR