Anti-PTEN Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAF822HU01
MSPP-PAF822HU1EA 280 USD
MSPP-PAF822HU1
Anti-PTEN Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Phosphatase And Tensin Homolog (PTEN), derived from recombinant PTEN(Thr2~Val403), is reactive with Human/Rat/Guinea pig/Dog/Pig/Bovine/Goat/Sheep/Horse/Chicken.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

PTEN acts as a tumor suppressor gene through the action of its phosphatase protein product. This phosphatase is involved in the regulation of the cell cycle, preventing cells from growing and dividing too rapidly.This gene was identified as a tumor suppressor that is mutated in a large number of cancers at high frequency. The protein encoded by this gene is a phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase. It contains a tensin like domain as well as a catalytic domain similar to that of the dual specificity protein tyrosine phosphatases. this protein preferentially dephosphorylates phosphoinositide substrates. It negatively regulates intracellular levels of phosphatidylinositol-3,4,5-trisphosphate in cells and functions as a tumor suppressor by negatively regulating Akt/PKB signaling pathway.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Guinea pig, Dog, Pig, Bovine, Goat, Sheep, Horse, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR