Anti-CRYl1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAF354HU02
MSPP-PAF354HU2EA 350 USD
MSPP-PAF354HU2
Anti-CRYl1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Crystallin Lambda 1 (CRYl1), derived from recombinant CRYl1 (Leu24~Tyr232), is reactive with Human/Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The enzyme requires NAD(H) as a coenzyme, and is inhibited by inorganic phosphate. A similar gene in the rabbit is thought to serve a structural role in the lens of the eye. Human CRYL1 encodes an enzyme-crystallin with a predicted length of 319 amino acids. The protein shares 83, 84, 85, 78, and 75% sequence identity with rabbit, mouse, rat, cow, and pig lambda-crystallins. It contains 2 domains characteristic of HCDH proteins: one in the C terminus and another, an NAD-binding domain, in the N terminus. Northern blot analysis of human tissues detected ubiquitous expression of a 1.5-Kb CRYL1 transcript, at highest levels in liver and kidney. Northern blot analysis and semiquantitative RT-PCR on 60 hepatocellular carcinoma samples showed that CRYL1 was downregulated in 58% of the samples.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: CRYl1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR