Anti-PANX1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAE788HU01
MSPP-PAE788HU1EA 350 USD
MSPP-PAE788HU1
Anti-PANX1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Pannexin 1 (PANX1), derived from recombinant PANX1 (Cys228~Asn411), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Pannexin 1 belongs to the innexin family. Innexin family members are the structural components of gap junctions. This protein and pannexin 2 are abundantly expressed in central nerve system (CNS) and are coexpressed in various neuronal populations. Studies in Xenopus oocytes suggest that this protein alone and in combination with pannexin 2 may form cell type-specific gap junctions with distinct properties. The deduced 48-kD protein contains 4 transmembrane domains and cytoplasmic N and C termini. Northern blot analysis detected Panx1 expression in rat eye, thyroid, prostate, kidney, liver, and central nervous system. In situ hybridization showed Pnx1 expression in several brain regions, including cortex, striatum, olfactory bulb, hippocampus, thalamus, and cerebellum.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PANX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR