Anti-IL11Ra Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAE771MU02
MSPP-PAE771MU2EA 360 USD
MSPP-PAE771MU2
Anti-IL11Ra Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Interleukin 11 Receptor Alpha (IL11Ra), derived from recombinant IL11Ra (Asn194~Gln329), is reactive with Mouse/Human/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

IL-11 receptor is a member of the hematopoietic cytokine receptor family. This particular receptor is very similar to ciliary neurotrophic factor, since both contain an extracellular region with a 2-domain structure composed of an immunoglobulin-like domain and a cytokine receptor-like domain. Alternative splicing has been observed at this locus and two variants, each encoding a distinct isoform, have been identified. This receptor was found to be a 422-amino acid protein containing a signal peptide followed by extracellular, transmembrane, and cytoplasmic domains. The extracellular region has a 2-domain structure homologous to those of the receptors for IL6R and ciliary neurotrophic factor (CNTFR), an immunoglobulin-like domain and a cytokine receptor-like domain.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IL11Ra
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR