Anti-IFI30 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAE762HU01
MSPP-PAE762HU1EA 350 USD
MSPP-PAE762HU1
Anti-IFI30 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Interferon Gamma Inducible Protein 30 (IFI30), derived from recombinant IFI30(Met1~Lys250), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Gamma-interferon-inducible lysosomal thiol reductase is a lysosomal thiol reductase that at low pH can reduce protein disulfide bonds. The enzyme is expressed constitutively in antigen-presenting cells and induced by gamma-interferon in other cell types. This enzyme has an important role in MHC class II-restricted antigen processing.,Arunachalam et al. (2000) described a lysosomal thiol reductase, which they called GILT, that was optimally active at low pH and capable of catalyzing disulfide bond reduction both in vivo and in vitro. They found that the GILT gene encodes a predicted 261-amino acid protein composed of a 37-amino acid signal peptide and a 224-amino acid proform.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IFI30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR