Anti-TUBb3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAE711HU01
MSPP-PAE711HU1EA 350 USD
MSPP-PAE711HU1
Anti-TUBb3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Tubulin Beta 3 (TUBb3), derived from recombinant TUBb3 (Tyr222~Glu438), is reactive with Human/Mouse/Rat/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

TUBB3 expression in several human cell lines, but not in a paclitaxel-resistant cell line. RNA interference revealed that HIF1A mediated hypoxia-induced TUBB3 expression. Chromatin immunoprecipitation analysis showed that HIF1A bound an HIF1A-binding site in the 5-prime flanking region of the TUBB3 gene. Methylation of an enhancer in the 3-prime flanking region abolished hypoxia-induced TUBB3 expression. Much lower Tubb3 levels were detected in all other adult mouse tissues examined except brain, where expression was even lower. Chromatin immunoprecipitation analysis showed that HIF1A bound an HIF1A-binding site in the 5-prime flanking region of the TUBB3 gene. Methylation of an enhancer in the 3-prime flanking region abolished hypoxia-induced TUBB3 expression.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: TUBb3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR