Anti-DEFa6 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAE134HU01
MSPP-PAE134HU1EA 350 USD
MSPP-PAE134HU1
Anti-DEFa6 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Defensin Alpha 6, Paneth Cell Specific (DEFa6), derived from recombinant DEFa6(Ala19~Cys99), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes appear to be clustered on chromosome 8.

Defensin Alpha 6 is highly expressed in the secretory granules of Paneth cells of the small intestine, and likely plays a role in host defense of human bowel.Paneth cells of the small intestine.Has very low antimicrobial activity against Gram-negative and Gram-positive bacteria. May protect cells against infection with HIV-1.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: DEFa6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR