Anti-PCDH15 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAE095MU01
MSPP-PAE095MU1EA 360 USD
MSPP-PAE095MU1
Anti-PCDH15 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Protocadherin 15 (PCDH15), derived from recombinant PCDH15 (Pro45~Phe270), is reactive with Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

PCSH15 encodes a variety of isoforms with 3 to 11 ectodomains, a transmembrane domain, and a C-terminal cytoplasmic domain (CD). The amino acid sequences of CD1, CD2, and CD3 are entirely different. PCDH15-CD1 showed a limited pattern of expression and was detected in human testis, retina, and cochlea. PCDH15-CD2 expression was present in the human heart, kidney, thymus, spleen, testis, retina and cochlea. PCDH15-CD3 was widely transcribed and detected in almost all of 18 tissues tested. PCDH15 expression in the inner and outer synaptic layers and the nerve fiber layer in human adult and fetal retinas. Additional reactivity in the region of the outer limiting membrane/photoreceptor cell inner segments was observed in adult but not fetal retina.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PCDH15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR