Anti-RORa Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAD947HU01
MSPP-PAD947HU1EA 350 USD
MSPP-PAD947HU1
Anti-RORa Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to RAR Related Orphan Receptor Alpha (RORa), derived from recombinant RORa(Gln164~Met401), is reactive with Human/Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

ROR-alpha), also known as NR1F1 (nuclear receptor subfamily 1, group F, member 1) is a nuclear receptor encoded by the RORA gene. The protein encoded by this gene is a member of the NR1 subfamily of nuclear hormone receptors. It can bind as a monomer or as a homodimer to hormone response elements upstream of several genes to enhance the expression of those genes. The specific functions of this protein are not known, but it has been shown to interact with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation, as well as with NM23-1, the product of a tumor metastasis suppressor candidate gene. Four transcript variants encoding different isoforms have been described for this gene.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: RORa
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR