Anti-CYP3A4 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAD299HU01
MSPP-PAD299HU1EA 350 USD
MSPP-PAD299HU1
Anti-CYP3A4 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Cytochrome P450 3A4 (CYP3A4), derived from recombinant CYP3A4 (Pro344~Asp497), is reactive with Human/Rat/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

CYP3A4is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. This enzyme is involved in the metabolism of approximately half the drugs which are are used today, including acetaminophen, codeine, cyclosporin A, diazepam and erythromycin. The enzyme also metabolizes some steroids and carcinogens. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Previously another CYP3A gene, CYP3A3, was thought to exist; however, it is now thought that this sequence represents a transcript variant of CYP3A4.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: CYP3A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR