Anti-GIF Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAD111MU01
MSPP-PAD111MU1EA 360 USD
MSPP-PAD111MU1
Anti-GIF Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Gastric Intrinsic Factor (GIF), derived from recombinant GIF (Asp31~Cys288), is reactive with Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Intrinsic factor is a glycoprotein produced by the parietal cells of the stomach. It is necessary for the absorption of vitamin B12 later on in the terminal ileum., Upon entry into the stomach, vitamin B12 becomes bound to haptocorrin (R factor), a glycoprotein. The resulting complex enters the duodenum, where pancreatic enzymes digest haptocorrin. In the less acidic environment of the small intestine, B12 can then bind to intrinsic factor. This new complex travels to the ileum, where special epithelial cells endocytose them. Inside the cell, B12 dissociate once again and bind to another protein, transcobalamin II. The new complex can exit the epithelial cells to enter the liver.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: GIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR