Anti-LRP1B Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAD103MU01
MSPP-PAD103MU1EA 360 USD
MSPP-PAD103MU1
Anti-LRP1B Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Low Density Lipoprotein Receptor Related Protein 1B (LRP1B), derived from recombinant LRP1B (Leu31~Lys194), is reactive with Human/Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

LRP1B belongs to the low density lipoprotein (LDL) receptor gene family. full-length LRP1B was unstable following transfection in Lrp1 null Chinese hamster ovary (CHO) cells. In order to characterize the ligands for LRP1B, generated a minireceptor of LRP1B that could be stably expressed in CHO cells. The minireceptor began with the EGF precursor-like repeat before the fourth cluster of ligand-binding repeats (domain IV) and contained ligand-binding domain IV, the transmembrane domain, and the cytoplasmic tail. Ligands for the minireceptor, including receptor-associated protein, urokinase plasminogen activator, tissue-type plasminogen activator, and plasminogen activator inhibitor-1, were bound, internalized, and degraded via their association with the minireceptor.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: LRP1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR