Anti-GPX4 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC994MU01
MSPP-PAC994MU1EA 360 USD
MSPP-PAC994MU1
Anti-GPX4 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Glutathione Peroxidase 4 (GPX4), derived from recombinant GPX4(Gly74~Leu197), is reactive with Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The antioxidant enzyme GPx4) belongs to the family of glutathione peroxidases, which consists of 8 known mammalian isoenzymes (GPx1-8). Gpx4 catalyzes the reduction of hydrogen peroxide, organic hydroperoxides, and lipid peroxides at the expense of reduced glutathione and functions in the protection of cells against oxidative stress. The oxidized form of glutathione (glutathione disulfide), which is generated during the reduction of hydroperoxides by GPx4, is recycled by glutathione reductase and NADPH/H+. GPx4 differs from the other GPx family members in terms of its monomeric structure, a less restricted dependence on glutathione as reducing substrate, and the ability to reduce lipid-hydroperoxides inside biological membranes.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: GPX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR