Anti-TSHR Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC825RA01
MSPP-PAC825RA1EA 380 USD
MSPP-PAC825RA1
Anti-TSHR Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Thyroid Stimulating Hormone Receptor (TSHR), derived from recombinant TSHR (Pro28~Gly245), is reactive with Rat/Human/Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The thyrotropin receptor is a protein responds to thyroid-stimulating hormone (also known as 'thyrotropin', and stimulates the production of thyroxine (T4) and triiodothyronine (T3). The TSH receptor is a member of the G-protein coupled receptor superfamily of integral membrane proteins.It is primarily found on the surface of the thyroid epithelial cells. It should not be confused with 'thyrotropin-releasing hormone' (TRH) or 'thyrotropin-releasing hormone receptor' (TRHR). The deduced 764-amino acid protein has a molecular mass of 86.8 kD and contains a signal peptide, 7 transmembrane regions, 5 potential glycosylation sites, and a short intracytoplasmic region. The TSHR cDNA encoded a functional receptor that activated adenylate cyclase in response to TSH.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: TSHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR