Anti-TP Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC822HU01
MSPP-PAC822HU1EA 350 USD
MSPP-PAC822HU1
Anti-TP Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Thromboxane Receptor (TP), derived from recombinant TP (Pro3~Ser324), is reactive with Human/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The TP) is a protein on the surface of cells in the endothelium of blood vessels and in the placenta which interacts with the eicosanoid lipid thromboxane. It is a G-protein coupled receptors coupled to the G protein Gq.The gene responsible for the thromboxane receptor, TBXA2R is found on chromosome 19 and spans 15 kilobases., Thromboxane is a potent stimulator for platelet aggregation and clot formation and also plays a role in vascular tone. The thromboxane receptor is a member of the family of G protein-coupled receptors. Mutation of the receptor can lead to a bleeding disorder.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: TP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR