Anti-RGN Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC765HU01
MSPP-PAC765HU1EA 350 USD
MSPP-PAC765HU1
Anti-RGN Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Regucalcin (RGN), derived from recombinant RGN (Gln65~Gly299), is reactive with Human/Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Regucalcinis a highly conserved, calcium-binding protein, that is preferentially expressed in the liver and kidney. It may have an important role in calcium homeostasis. Studies in rat indicate that this protein may also play a role in aging, as it shows age-associated down-regulation. This gene is part of a gene cluster on chromosome Xp11.3-Xp11.23. Alternative splicing results in two transcript variants having different 5' UTRs, but encoding the same protein. Regucalcin is a proposed name for a calcium-binding protein that was discovered in 1978. Regucalcin differs from calmodulin and other Ca²⁺-related proteins as it does not contain an EF-hand motif of Ca²⁺-binding domain. It may regulate the effect of Ca²⁺ on liver cell functions.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: RGN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR