Anti-PAH Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC750HU01
MSPP-PAC750HU1EA 350 USD
MSPP-PAC750HU1
Anti-PAH Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Phenylalanine Hydroxylase (PAH), derived from recombinant PAH (Met1~Gln226), is reactive with Human/Mouse/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Phenylalanine hydroxylase is the rate-limiting enzyme of the metabolic pathway which degrades excess phenylalanine. The other substrates in the reaction are molecular oxygen and tetrahydrobiopterin. Tetrahydrobiopterin is a member of the group of redox biochemicals known as pteridines., PAH is the gene that encodes for phenylalanine hydroxylase. It was the research on phenylalanine hydroxylase by Seymour Kaufman that led to the discovery of tetrahydrobiopterin as a biological cofactor. Phenylalanine hydroxylase is a tetramer composed of four monomers, that is, composed of 4 identical subunits. Each subunit is in turn composed of three domains, a regulatory domain, a catalytic domain, and a tetramerization domain.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PAH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR