Anti-MSRA Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC629HU01
MSPP-PAC629HU1EA 350 USD
MSPP-PAC629HU1
Anti-MSRA Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Methionine Sulfoxide Reductase A (MSRA), derived from recombinant MSRA (Ala27~Lys235), is reactive with Human/Rat/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Peptide methionine sulfoxide reductase is ubiquitous and highly conserved. It carries out the enzymatic reduction of methionine sulfoxide to methionine. Human and animal studies have shown the highest levels of expression in kidney and nervous tissue. MSRA mRNA is highly expressed in rat and human kidney, but to a lesser extent in other rat tissues. The highest specific activity of the mammalian MSRA enzyme was found in cell-free extracts prepared from rat kidney and human neutrophils. Neutrophils and macrophages are known to produce reactive oxygen species during a respiratory burst that kill invading bacteria during phagocytosis. It is suggested that MSRA functions to repair oxidative damage to proteins that may occur during oxidative bursts or other situations where reactive oxygen radicals exist.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MSRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR