Anti-MARCO Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC614MU01
MSPP-PAC614MU1EA 360 USD
MSPP-PAC614MU1
Anti-MARCO Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Macrophage Receptor With Collagenous Structure (MARCO), derived from recombinant MARCO(Gly366~Ser518), is reactive with Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Macrophage receptor MARCO is a member of the class A scavenger receptor family and is part of the innate antimicrobial immune system. The protein may bind both Gram-negative and Gram-positive bacteria via an extracellular, C-terminal, scavenger receptor cysteine-rich (SRCR) domain. In addition to short cytoplasmic and transmembrane domains, there is an extracellular spacer domain and a long, extracellular collagenous domain. The protein may form a trimeric molecule by the association of the collagenous domains of three identical polypeptide chains. Alveolar macrophages (AMs) express the class A scavenger receptor macrophage receptor with collagenous structure (MARCO), but its role in vivo in lung defense against bacteria and environmental particles has not been studied.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MARCO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR