Anti-KISS1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC559MU01
MSPP-PAC559MU1EA 360 USD
MSPP-PAC559MU1
Anti-KISS1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Kisspeptin 1 (KISS1), derived from recombinant KISS1(Met1~Gly126), is reactive with Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Kisspeptin is a G-protein coupled receptor ligand for GPR54. Kiss1 was originally identified as a human metastasis suppressor gene that has the ability to suppress melanoma and breast cancer metastasis. It is recently become clear that kisspeptin-GPR54 signaling has an important role in initiating GnRH secretion at puberty, the extent of which is an area of ongoing research.Kisspeptin is a metastasis suppressor of melanomas and breast cancers without affecting tumorigenicity. The encoded protein may function to inhibit chemotaxis and invasion, attenuating metastasis in malignant melanomas. Studies suggest a putative role in the regulation of events downstream of cell-matrix adhesion, perhaps involving cytoskeletal reorganization. A polymorphism in the terminal exon of this mRNA results in two protein isoforms.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: KISS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR