Anti-FXN Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC493HU01
MSPP-PAC493HU1EA 350 USD
MSPP-PAC493HU1
Anti-FXN Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Frataxin (FXN), derived from recombinant FXN (Ser57~Leu198), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Frataxin is a small protein, localized to the mitochondrion. The function of frataxin is not entirely clear, but it seems to be involved in assembly of iron-sulfur clusters. It has been proposed to act as either an iron Chaperone (protein) or an iron storage protein. Reduced expression of frataxin is the cause of Friedrich's ataxia (FRDA), a lethal neurodegenerative disease. This nuclear gene encodes a mitochondrial protein which belongs to FRATAXIN family. The protein functions in regulating mitochondrial iron transport and respiration. The expansion of intronic trinucleotide repeat GAA results in Friedreich ataxia. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: FXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR