Anti-ERN1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC460HU01
MSPP-PAC460HU1EA 350 USD
MSPP-PAC460HU1
Anti-ERN1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Endoplasmic Reticulum To Nucleus Signalling 1 (ERN1), derived from recombinant ERN1(Phe571~Phe832), is reactive with Human/Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Serine/threonine-protein kinase/endoribonuclease IRE1 is the ER to nucleus signalling 1 protein, a human homologue of the yeast Ire1 gene product. This protein possesses intrinsic kinase activity and an endoribonuclease activity and it is important in altering gene expression as a response to endoplasmic reticulum-based stress signals. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. Senses unfolded proteins in the lumen of the endoplasmic reticulum via its N-terminal domain which leads to enzyme auto- activation. The active endoribonuclease domain splices XBP1 mRNA to generate a new C-terminus, converting it into a potent unfolded-protein response transcriptional activator and triggering growth arrest and apoptosis.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: ERN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR