Anti-COL16 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC136HU01
MSPP-PAC136HU1EA 350 USD
MSPP-PAC136HU1
Anti-COL16 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Collagen Type XVI (COL16), derived from recombinant COL16(Val1434~Gly1604), is reactive with Human/Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

XVI collagen is a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Members of this collagen family are found in association with fibril-forming collagens such as type I and II, and serve to maintain the integrity of the extracellular matrix. High levels of type XVI collagen have been found in fibroblasts and keratinocytes, and in smooth muscle and amnion. The remaining collagens belong to the latter class with a common feature being the presence of imperfections in the gly-Xaa-Xaa repeating pattern. Within the latter class, collagens type IX, XII, and XIV form a subgroup called the FACIT collagens (for 'fibril-associated collagens with interrupted triple helices'). These collagens are associated with type I or II collagen fibrils and play a role in interaction of these fibrils with other matrix components.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: COL16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR