Anti-GDF5 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC110HU01
MSPP-PAC110HU1EA 340 USD
MSPP-PAC110HU1
Anti-GDF5 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Growth Differentiation Factor 5 (GDF5), derived from recombinant GDF5 (Ala382~Arg501), is reactive with Human/Mouse/Rat/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Growth/differentiation factor 5 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues., Growth differentiation factor 5 (GDF5) is a protein belonging to the transforming growth factor beta superfamily that is expressed in the developing central nervous system, and has a role in skeletal and joint development.It also increases the survival of neurones that respond to a neurotransmitter called dopamine, and is a potential therapeutic molecule associated with Parkinson's disease.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: GDF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR