Anti-IL35 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC008HU01
MSPP-PAC08HU01EA 350 USD
MSPP-PAC08HU01
Anti-IL35 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Interleukin 35 (IL35), derived from recombinant IL35 (TheIL35complexcomposedofIL27Bsubunit (Arg21~Lys229)andthematureformofhumanIL12A (Arg23~Ser219)linkedbyapolypeptidelinker (GGGGS)3), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Interleukin 35 (IL-35) is an IL-12 family cytokine that is produced by regulatory, but not effector, T cells and plays a role in immune suppression. It is a dimeric protein composed of IL-12alpha; and IL-27beta; chains, which are encoded by two separate genes called IL12A and EBI3, respectively. Secreted by regulatory T cells (Tregs), IL-35 suppresses inflammatory responses of immune cells. Studies in mice show the absence of either IL-35 chain from regulatory Tregs reduces the cells ability to suppress inflammation; this has been observed during cell culture experiments and using an experimental model for inflammatory bowel disease. To produce its suppressive effects, IL-35 has selective activities on different T cell subsets; it induces proliferation of Treg cell populations but reduces activity of Th17 cell populations.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IL35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR