Anti-RANk Rabbit Polyclonal Antibody

Supplier: 0000027876
PAC057HU01
MSPP-PAC057HU1EA 350 USD
MSPP-PAC057HU1
Anti-RANk Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Receptor Activator Of Nuclear Factor Kappa B (RANk), derived from recombinant RANk (Val330~Lys615), is reactive with Human/Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

RANk is a member of the TNF-receptor superfamily. This receptors can interact with various TRAF family proteins, through which this receptor induces the activation of NF-kappa B and MAPK8/JNK. This receptor and its ligand are important regulators of the interaction between T cells and dendritic cells. This receptor is also an essential mediator for osteoclast and lymph node development. Studies of the mouse counterpart suggest that this receptor directly mediates the osteoprotegerin ligand (OPGL)-induced osteoclastogenesis in osteoclast precursor cells. Northern blot analysis indicated that the 4.5-kb human RANK mRNA is expressed ubiquitously. In some human cell lines, the authors detected additional transcripts that were derived from the use of alternative polyadenylation signals in the RANK gene.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: RANk
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR