Anti-MMP14 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC056HU02
MSPP-PAC056HU2EA 350 USD
MSPP-PAC056HU2
Anti-MMP14 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Matrix Metalloproteinase 14 (MMP14), derived from recombinant MMP14 (Asn322~Ala541), is reactive with Human/Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

MMP14, also known as MT1-MMP, is a member of membrane type (MT) MMPs. It can degrade various components of the extracellular matrix such as type I collagen and activate pro-MMP-2 and process cell adhesion molecules such as CD44 and integrin alpha V. It is also essential for pericellular collagenolysis and modeling of skeletal and extra skeletal connective tissues during development. MMP14 plays a important role in many physiological and pathological processes such as angiogenesis and tumor invasion. This protein is mainly expressed in stromal cells of colon, breast, head and neck. There are four domains for native MMP14, a pro domain containing the furin cleavage site, a catalytic domain containing the zinc-binding site, a hemopexin-like domain and a transmembrane domain.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MMP14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR