Anti-rT3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC022GE01
MSPP-PAC022GE1EA 398 USD
MSPP-PAC022GE1
Anti-rT3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Reverse Triiodothyronine (rT3), derived from OVA conjugated rT3, is reactive with General species.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Reverse triiodothyronine is a molecule which is an isomer of triiodothyronine (T3). It is derived from thyroxine (T4) through the action of deiodinase. rT3, unlike T3, does not stimulate thyroid hormone receptors. However, rT3 nonetheless binds to these receptors, thereby blocking the action of T3. Under stress conditions, the adrenal glands produce excess amounts of cortisol. Cortisol inhibits the conversion of T4 to T3, thus shunting T4 conversion from T3 towards rT3.,Consequently, there is a widespread shutdown in T3 binding across the body. This condition is termed Reverse T3 Dominance. It results in reduced body temperature, which slows the action of many enzymes, leading to a clinical syndrome, Multiple Enzyme Dysfunction, which produces the effects seen in hypothyroidism. Effects include fatigue, headache, migraine, PMS, irritability, fluid retention, anxiety and panic.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: rT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: General species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR