Anti-CYP17A1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAC016RA01
MSPP-PAC016RA1EA 380 USD
MSPP-PAC016RA1
Anti-CYP17A1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Cytochrome P450 17A1 (CYP17A1), derived from recombinant CYP17A1(Thr201~Thr507), is reactive with Human/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

CYP17A1 (also known as 17 alpha-hydroxylase/17,20 lyase/17,20 desmolase) is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. It has both 17alpha-hydroxylase and 17,20-lyase activities and is a key enzyme in the steroidogenic pathway that produces progestins, mineralocorticoids, glucocorticoids, androgens, and estrogens. Mutations in this gene are associated with isolated steroid-17 alpha-hydroxylase deficiency, 17-alpha-hydroxylase/17,20-lyase deficiency, pseudohermaphroditism, and adrenal hyperplasia.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: CYP17A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR