Anti-MAPT Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB983MU01
MSPP-PAB983MU1EA 342 USD
MSPP-PAB983MU1
Anti-MAPT Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Tau Protein (MAPT), derived from recombinant MAPT (Ala92~Val400), is reactive with Mouse/Rat/Pig.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Tau protein is a highly soluble microtubule-associated protein (MAP). In humans, these proteins are mostly found in neurons compared to non-neuronal cells. One of tau's main functions is to modulate the stability of axonal microtubules. Tau is not present in dendrites and is active primarily in the distal portions of axons where it provides microtubule stabilization but also flexibility as needed. This contrasts with STOP proteins in the proximal portions of axons which essentially lock down the microtubules and MAP2 that stabilizes microtubules in dendrites. The tau gene locates on chromosome 17q21, containing 16 exons. The major tau protein in the human brain is encoded by 11 exons. Exon 2, 3 and 10 are alternative spliced, allowing six combinations (2-3-10-; 2+3-10-; 2+3+10-; 2-3-10+; 2+3-10+; 2+3+10+).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR