Anti-F11 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB834MU01
MSPP-PAB834MU1EA 360 USD
MSPP-PAB834MU1
Anti-F11 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Coagulation Factor XI (F11), derived from recombinant F11 (Pro157~Arg389), is reactive with Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Factor XI is the zymogen form of factor XIa, one of the enzymes of the coagulation cascade. Like many other coagulation factors, it is a serine protease. Factor XI (FXI) is produced by the liver and circulates as a homo-dimer in its inactive form. The plasma half-life of FXI is approximately 52 hours. The zymogen factor is activated into factor XIa by factor XIIa (FXIIa), thrombin, and it is also autocatalytic, and FXI is a member of the 'contact pathway' due to activation by FXIIa (with includes HMWK, prekallikrein, factor XII, factor XI and factor IX). Factor XIa activates factor IX by selectively cleaving arg-ala and arg-val peptide bonds. Factor IXa, in turn, activates factor X. Inhibitors of factor XIa include protein Z-dependent protease inhibitor (ZPI), which is independent of protein Z.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: F11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR