Anti-S100A9 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB793HU01
MSPP-PAB793HU1EA 332 USD
MSPP-PAB793HU1
Anti-S100A9 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to S100 Calcium Binding Protein A9 (S100A9), derived from recombinant S100A9 (Met1~Pro114), is reactive with Human/Mouse/Rat/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

S100 calcium binding protein A9 has been implicated in the abnormal differentiation of myeloid cells in the stroma of cancer. This contributes to creating an overall immunosuppressive microenvironemnt that may contribute to the inability of a protective or therapeutic cellular immune response to be generated by the tumor-bearing host. Outside of malignancy, S100A9 in association with its dimerization partner, S100A8 (MRP8 or calgranulin A) signals for lymphocyte recruitment in sites of inflammation. S100A9/A8 (synonyma: Calgranulin A/B; Calprotectin) are also regarded as marker proteins for a number of inflammatory diseases in humans, especially in rheumatoid arthritis.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: S100A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR