Anti-RNASE3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB758HU01
MSPP-PAB758HU1EA 332 USD
MSPP-PAB758HU1
Anti-RNASE3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Ribonuclease A3 (RNASE3), derived from recombinant RNASE3(Pro30~Val154), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Eosinophil cationic protein, like eosinophil-derived neurotoxin, is localized to the granule matrix of the eosinophil. Both proteins possess neurotoxic, helminthotoxic, and ribonucleolytic activities. The cDNA sequences of the genes encoding these 2 proteins and the deduced amino acid sequences indicate that the genes belong to the ribonuclease gene superfamily. Hamann et al. (1990) demonstrated remarkable similarities of eosinophil-derived neurotoxin and eosinophil cationic protein, symbolized RNS2 and RNS3, respectively, in noncoding sequences, introns, and flanking regions, as well as the previously known coding regions. A single intron in the 5-prime untranslated region and an intronless coding region appear to be features common to many members of the RNase gene superfamily.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: RNASE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR