Anti-OIT3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB729HU01
MSPP-PAB729HU1EA 332 USD
MSPP-PAB729HU1
Anti-OIT3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Oncoprotein Induced Transcript 3 (OIT3), derived from recombinant OIT3(Val297~Arg506), is reactive with Human/Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

OIT3 contains one EGF-like domain and one ZP domain. It may be involved in hepatocellular function and development. OIT3 is liver-specific and is expressed only in the hepatocytes, down-regulated in hepatocellular carcinoma (HCC) and HCC cell lines. LZP contains an N-terminal signal peptide, followed by 3 epidermal growth factor (EGF)-like domains and a zona pellucida (ZP) domain in its C-terminal region. LZP also contains several N-glycosylation sites. The 546-amino acid mouse Lzp protein shares 89% identity with human LZP. Northern blot analysis of several human tissues detected a 2.4-kb transcript only in liver. Northern blot analysis of whole mouse embryos detected 2 Lzp variants that differed in the length of the UTR and the poly(A) signal used.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: OIT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR