Anti-CD163 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB726HU01
MSPP-PAB726HU1EA 332 USD
MSPP-PAB726HU1
Anti-CD163 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Cluster Of Differentiation (CD163), derived from recombinant CD163(Leu719~Thr1029), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

CD163, a hemoglobin scavenger receptor, is expressed in monocytes and macrophages.Acute phase-regulated receptor involved in clearance and endocytosis of hemoglobin/haptoglobin c by macrophages and may thereby protect tissues from free hemoglobin-mediated oxidative damage.,CD163 may play a role in the uptake and recycling of iron, via endocytosis of hemoglobin/haptoglobin and subsequent breakdown of heme. It binds hemoglobin/haptoglobin complexes in a calcium-dependent and pH-dependent manner. It exhibits a higher affinity for complexes of hemoglobin and multimeric haptoglobin of HP*1F phenotype than for complexes of hemoglobin and dimeric haptoglobin of HP*1S phenotype.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: CD163
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR