Anti-MBP Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB650MU01
MSPP-PAB650MU1EA 342 USD
MSPP-PAB650MU1
Anti-MBP Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Major Basic Protein (MBP), derived from recombinant MBP (Met100~Lys215), is reactive with Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Major basic protein The protein encoded by this gene is the predominant constituent of the crystalline core of the eosinophil granule. High levels of the proform of this protein are also present in placenta and pregnancy serum, where it exists as a complex with several other proteins including pregnancy-associated plasma protein A (PAPPA), angiotensinogen (AGT), and C3dg. This protein may be involved in antiparasitic defense mechanisms as a cytotoxin and helminthotoxin, and in immune hypersensitivity reactions. It is directly implicated in epithelial cell damage, exfoliation, and bronchospasm in allergic diseases.PRG2 is a 117-residue protein that predominates in eosinophil granules. It is a potent enzyme against helminths and is toxic towards bacteria and mammalian cells in vitro.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR