Anti-CD164 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB594MU01
MSPP-PAB594MU1EA 342 USD
MSPP-PAB594MU1
Anti-CD164 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Cluster Of Differentiation 164 (CD164), derived from recombinant CD164 (Gln24~Asp162), is reactive with Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

CD164 is a transmembrane isoform of the mucin-like glycoprotein MGC-24. MGC-24 is a 24 kDa protein rich in hydroxyl amino acids and cysteine, and lack repeating motifs identified originally in human gastric carcinoma cells as a protein binding peanut agglutinin. High levels of MGC-24 mRNA are expressed in normal tissues of the colon, small intestine and thyroid, and in some nude mouse-transplanted colorectal and pancreatic adenocarcinomas. Kurosawa et al (1999) have reported that in the mouse only the transmembrane form is expressed in many organs. CD164 is expressed by human CD34(+) hematopoietic progenitor cells. The subpopulations of CD34(+) cells expressing CD164 include the majority of CFU-GM, BFU-E, and hierarchically more primitive precursors (pre-CFU)..

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: CD164
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR