Anti-HBEGF Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAB479HU01
MSPP-PAB479HU1EA 280 USD
MSPP-PAB479HU1
Anti-HBEGF Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF), derived from recombinant HBEGF (Val21~Thr160), is reactive with Human/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Heparin-binding EGF-like growth factor (HB-EGF) is a member of the EGF family of proteins. It has been shown to play a role in wound healing, cardiac hypertrophy and heart development and function. Diphtheria toxin inhibits protein synthesis in eukaryotic cells by catalyzing inactivation of elongation factor-2 (EF2). Entry of toxin into cells is dependent on binding to the cell surface.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: HBEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR