Anti-TH Rabbit Polyclonal Antibody

Supplier: 0000027876
PAB438MU01
MSPP-PAB438MU1EA 342 USD
MSPP-PAB438MU1
Anti-TH Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Tyrosine Hydroxylase (TH), derived from recombinant TH(Phe34~Leu301), is reactive with Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Tyrosine hydroxylase or tyrosine 3-monooxygenase is the enzyme responsible for catalyzing the conversion of the amino acid L-tyrosine to dihydroxyphenylalanine (DOPA). DOPA is a precursor for dopamine which in turn is a precursor for norepinephrine (noradrenaline) and epinephrine (adrenaline). In humans, tyrosine hydroxylase is encoded by the TH gene. The enzyme, an oxygenase, is found in the cytosol of all cells containing catecholamines. This initial reaction is the rate limiting step in the production of catecholamines. The enzyme is highly specific, not accepting indole derivatives - which is unusual as many other enzymes involved in the production of catecholamines do.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR