Anti-SEMA4D Rabbit Polyclonal Antibody

Supplier: 0000027876
PAB430MU01
MSPP-PAB430MU1EA 342 USD
MSPP-PAB430MU1
Anti-SEMA4D Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Semaphorin 4D (SEMA4D), derived from recombinant SEMA4D (Phe502~Ser636), is reactive with Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

CD100 was first identified as a cell surface protein of resting T cells; previous studies had shown that it was involved in lymphocyte activation. Hall et al., (1996) cloned CD100 from a cDNA library of phytohemagglutinin-activated human T cells. Sequence analysis showed that CD100 encodes an 863-amino acid polypeptide containing a secretory signal sequence, a single transmembrane domain, an immunoglobulin (Ig)-like domain, and a conserved 500-amino acid sema domain. Thus, Hall et al., (1996) stated that CD100 is a member of the semaphorin family and the first semaphorin believed to be involved in the immune system. See SEMA3F and SEMA3B. Northern blot analysis revealed that CD100 is expressed primarily as a 4.5-kb message in a variety of tissues, both hematopoietic and nonhematopoietic.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: SEMA4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR